siRNA Details
| Gene Symbol: Wdr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175552.4 | NP_780761.1 | 3, 4 | 467 | ||
| GenBank mRNA | AK043557.1 | - | 3, 4 | 400 | ||
| AK050738.1 | - | 3, 4 | 435 | |||
| AK050797.1 | - | 3, 4 | 438 | |||
| BC063100.1 | - | 3, 4 | 453 | 6332749 | ||

Wdr3
Genomic Map