siRNA Details
| Gene Symbol: Lrig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025067.1 | NP_001020238.1 | 6 | 881 | ||
| GenBank mRNA | AK036116.1 | - | 6 | 882 | ||
| AK155089.1 | - | 6 | 944 | |||
| AK158022.1 | - | 7 | 898 | |||
| BC094228.1 | - | 6 | 818 | 30362610 | ||
| BC138422.1 | - | 6 | 792 | 8861442 | ||

Lrig2
Genomic Map