siRNA Details
| Gene Symbol: Lrig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025067.1 | NP_001020238.1 | 14 | 2172 | ||
| GenBank mRNA | AK028527.1 | - | 4 | 789 | ||
| AK036116.1 | - | 14 | 2174 | |||
| AK155089.1 | - | 13 | 3513 | |||
| AK173030.1 | - | 4 | 1070 | |||
| BC094228.1 | - | 14 | 2109 | 30362610 | ||
| BC138422.1 | - | 14 | 2083 | 8861442 | ||

Lrig2
Genomic Map