siRNA Details
Gene Symbol: Clspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175554.4 | NP_780763.3 | 19 | 3295 | ||
GenBank mRNA | AY324187.1 | - | 5 | 450 | ||
BC050848.1 | - | 19 | 3250 | 6507692 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175554.4 | NP_780763.3 | 19 | 3295 | ||
GenBank mRNA | AY324187.1 | - | 5 | 450 | ||
BC050848.1 | - | 19 | 3250 | 6507692 |