siRNA Details
| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175554.4 | NP_780763.3 | 7 | 1070 | ||
| GenBank mRNA | AK053847.1 | - | 7 | 1085 | ||
| AK135394.1 | - | 7 | 1049 | |||
| BC050848.1 | - | 7 | 1025 | 6507692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clspn| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175554.4 | NP_780763.3 | 7 | 1070 | ||
| GenBank mRNA | AK053847.1 | - | 7 | 1085 | ||
| AK135394.1 | - | 7 | 1049 | |||
| BC050848.1 | - | 7 | 1025 | 6507692 | ||