siRNA Details
| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175554.4 | NP_780763.3 | 20 | 3394 | ||
| GenBank mRNA | AK161785.1 | - | 2 | 88 | ||
| AY324187.1 | - | 6 | 549 | |||
| BC050848.1 | - | 20 | 3349 | 6507692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clspn| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175554.4 | NP_780763.3 | 20 | 3394 | ||
| GenBank mRNA | AK161785.1 | - | 2 | 88 | ||
| AY324187.1 | - | 6 | 549 | |||
| BC050848.1 | - | 20 | 3349 | 6507692 | ||