siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031393.2 | NP_113570.2 | 9, 10 | 1070 | ||
| GenBank mRNA | AB050741.1 | - | 9, 10 | 1046 | ||
| AK078868.1 | - | 9, 10 | 1059 | |||
| AK131162.1 | - | 9, 10 | 1027 | |||
| AK154176.1 | - | 9, 10 | 1063 | |||

Sytl1
Genomic Map