siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031393.2 | NP_113570.2 | 3 | 455 | ||
| GenBank mRNA | AB050741.1 | - | 3 | 434 | ||
| AK078868.1 | - | 3 | 443 | |||
| AK131162.1 | - | 3 | 412 | |||
| AK154176.1 | - | 3 | 450 | |||
| BC005623.1 | - | 3 | 455 | 3155535 | ||

Sytl1
Genomic Map