siRNA Details
| Gene Symbol: Chd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081376.1 | NP_001074845.1 | 14 | 2434 | ||
| NM_029216.2 | NP_083492.2 | 14 | 2434 | |||
| GenBank mRNA | AK147563.1 | - | 14 | 2435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chd5| Gene Symbol: Chd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081376.1 | NP_001074845.1 | 14 | 2434 | ||
| NM_029216.2 | NP_083492.2 | 14 | 2434 | |||
| GenBank mRNA | AK147563.1 | - | 14 | 2435 | ||