siRNA Details
| Gene Symbol: Rbm48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172991.4 | NP_766579.3 | 4 | 1457 | ||
| GenBank mRNA | AK047811.1 | - | 4 | 989 | ||
| BC029549.1 | - | 4 | 1321 | 5012510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm48| Gene Symbol: Rbm48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172991.4 | NP_766579.3 | 4 | 1457 | ||
| GenBank mRNA | AK047811.1 | - | 4 | 989 | ||
| BC029549.1 | - | 4 | 1321 | 5012510 | ||