siRNA Details
Gene Symbol: TAS2R38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176817.4 | NP_789787.4 | 1 | 776 | ||
GenBank mRNA | BC104933.1 | - | 1 | 776 | 8143936 | |
BC104937.1 | - | 1 | 776 | 8143940 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAS2R38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176817.4 | NP_789787.4 | 1 | 776 | ||
GenBank mRNA | BC104933.1 | - | 1 | 776 | 8143936 | |
BC104937.1 | - | 1 | 776 | 8143940 |