siRNA Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172993.3 | NP_766581.2 | 11 | 1313 | ||
| GenBank mRNA | AK084205.1 | - | 11 | 1260 | ||
| AK146353.1 | - | 11 | 1277 | |||
| AK149921.1 | - | 11 | 1260 | |||
| AK154918.1 | - | 11 | 1260 | |||
| AK173267.1 | - | 11 | 1288 | |||
| BC021520.1 | - | 9 | 1107 | |||
| BC066136.1 | - | 10 | 1066 | |||
| BC089160.1 | - | 11 | 1235 | 5390706 | ||

Zfp512
Genomic Map