siRNA Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172994.2 | NP_766582.1 | 4 | 612 | ||
| GenBank mRNA | AK032023.1 | - | 4 | 515 | ||
| AK049830.1 | - | 4 | 498 | |||
| AY348866.1 | - | 4 | 614 | |||
| BC059811.1 | - | 4 | 612 | 6849535 | ||

Ppp2r2c
Genomic Map