siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040106.2 | NP_001035195.1 | 20 | 3296 | ||
| NM_177762.6 | NP_808430.2 | 19 | 3053 | |||
| GenBank mRNA | AK129272.1 | - | 10 | 1514 | ||
| AK147363.1 | - | 20 | 3297 | |||
| AK166107.1 | - | 1 | 1079 | |||
| BC043125.1 | - | 9 | 1102 | |||

Aak1
Genomic Map