siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040106.2 | NP_001035195.1 | 7 | 1362 | ||
| NM_177762.6 | NP_808430.2 | 7 | 1362 | |||
| GenBank mRNA | AK147363.1 | - | 7 | 1363 | ||
| AK158879.1 | - | 7 | 1252 | |||
| BC141176.1 | - | 7 | 1118 | 9055788 | ||

Aak1
Genomic Map