siRNA Details
| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109749.1 | NP_001103219.1 | 18 | 2386 | ||
| NM_001109751.1 | NP_001103221.1 | 17 | 2474 | |||
| NM_173004.3 | NP_766592.2 | 18 | 2386 | |||
| GenBank mRNA | AK036262.1 | - | 18 | 2356 | ||
| AK044694.1 | - | 8 | 1133 | |||
| AK173340.1 | - | 11 | 1505 | |||
| BC046975.1 | - | 15 | 2008 | |||
| BC115766.1 | - | 18 | 2276 | 40060288 | ||

Cntn4
Genomic Map