siRNA Details
| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199311.2 | NP_955015.1 | 3 | 402 | ||
| GenBank mRNA | AY397675.1 | - | 3 | 337 | ||
| BC049354.1 | - | 3 | 392 | 6310008 | ||
| CK626014.1 | - | 3 | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4a1| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199311.2 | NP_955015.1 | 3 | 402 | ||
| GenBank mRNA | AY397675.1 | - | 3 | 337 | ||
| BC049354.1 | - | 3 | 392 | 6310008 | ||
| CK626014.1 | - | 3 | 402 | |||