siRNA Details
| Gene Symbol: Olfr1349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207136.1 | NP_997019.1 | 1 | 769 | ||
| GenBank mRNA | BC147396.1 | - | 1 | 791 | 9056466 | |
| BC147397.1 | - | 1 | 791 | 9056451 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1349| Gene Symbol: Olfr1349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207136.1 | NP_997019.1 | 1 | 769 | ||
| GenBank mRNA | BC147396.1 | - | 1 | 791 | 9056466 | |
| BC147397.1 | - | 1 | 791 | 9056451 | ||