siRNA Details
Gene Symbol: Olfr1349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207136.1 | NP_997019.1 | 1 | 157 | ||
GenBank mRNA | BC147396.1 | - | 1 | 179 | 9056466 | |
BC147397.1 | - | 1 | 179 | 9056451 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207136.1 | NP_997019.1 | 1 | 157 | ||
GenBank mRNA | BC147396.1 | - | 1 | 179 | 9056466 | |
BC147397.1 | - | 1 | 179 | 9056451 |