siRNA Details
| Gene Symbol: Zfp446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168561.1 | NP_001162033.1 | 4 | 836 | ||
| NM_001168562.1 | NP_001162034.1 | 5 | 1061 | |||
| NM_175558.4 | NP_780767.1 | 5 | 955 | |||
| GenBank mRNA | AK041761.1 | - | 5 | 957 | ||
| AK138378.1 | - | 3 | 1254 | |||
| AK142766.1 | - | 4 | 1450 | |||
| BC048909.1 | - | 5 | 897 | |||
| BC117850.1 | - | 5 | 1068 | |||
| BC117851.1 | - | 4 | 838 | 40093910 | ||

Zfp446
Genomic Map