siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 8 | 1098 | ||
| GenBank mRNA | AF212319.1 | - | 8 | 1069 | ||
| AK088110.1 | - | 8 | 1070 | |||
| AK145753.1 | - | 8 | 1043 | |||
| AK146564.1 | - | 8 | 1184 | |||
| AK161640.1 | - | 8 | 1082 | |||
| AK169395.1 | - | 8 | 1095 | |||
| AK177496.1 | - | 2 | 83 | |||
| BC060030.1 | - | 8 | 1086 | 6515657 | ||

Idh2
Genomic Map