siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 6 | 783 | ||
| GenBank mRNA | AF212319.1 | - | 6 | 754 | ||
| AK088110.1 | - | 6 | 755 | |||
| AK145753.1 | - | 6 | 728 | |||
| AK146564.1 | - | 6 | 869 | |||
| AK161640.1 | - | 6 | 767 | |||
| AK169395.1 | - | 6 | 780 | |||
| BC060030.1 | - | 6 | 771 | 6515657 | ||

Idh2
Genomic Map