siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177765.3 | NP_808433.2 | 15 | 2573 | ||
| GenBank mRNA | AK029565.1 | - | 15 | 2572 | ||
| AM690757.1 | - | 14 | 2274 | |||
| BC145749.1 | - | 14 | 2437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll13| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177765.3 | NP_808433.2 | 15 | 2573 | ||
| GenBank mRNA | AK029565.1 | - | 15 | 2572 | ||
| AM690757.1 | - | 14 | 2274 | |||
| BC145749.1 | - | 14 | 2437 | |||