siRNA Details
| Gene Symbol: Nup98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287164.1 | NP_001274093.1 | 23 | 3319 | ||
| NM_001287165.1 | NP_001274094.1 | 23 | 3268 | |||
| NM_022979.2 | NP_075355.1 | 23 | 3319 | |||
| GenBank mRNA | BC039912.1 | - | 8 | 1254 | ||
| BC050911.1 | - | 23 | 3273 | 6417631 | ||
| BC057608.1 | - | 23 | 3273 | 6417631 | ||
| BC078630.1 | - | 9 | 1322 | |||
| BC099600.1 | - | 4 | 477 | |||
| BC112912.1 | - | 23 | 3273 | 5697919 | ||
| BC137975.1 | - | 22 | 3127 | 8860994 | ||
| BC145071.1 | - | 22 | 3127 | 9053619 | ||

Nup98
Genomic Map