siRNA Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173446.2 | NP_775622.1 | 3 | 1160 | ||
| GenBank mRNA | AK032202.1 | - | 3 | 1160 | ||
| AK075681.1 | - | 3 | 291 | |||
| AK162130.1 | - | 3 | 697 | |||
| BC117535.1 | - | 4 | 1204 | 40106181 | ||

Fam155a
Genomic Map