siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 18, 19 | 2874 | ||
| XM_011515162.1 | XP_011513464.1 | 17, 18 | 2837 | |||
| GenBank mRNA | AB209744.1 | - | 18, 19 | 2828 | ||
| AF053944.1 | - | 18, 19 | 2708 | |||
| AK090931.1 | - | 5, 6 | 966 | |||
| AK127541.1 | - | 5, 6 | 1362 | |||
| AK296001.1 | - | 4, 5 | 868 | |||
| BC038588.1 | - | 18, 19 | 2874 | 5764769 | ||
| D86479.1 | - | 12, 13 | 1630 | |||

AEBP1
Genomic Map