siRNA Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044744.1 | NP_001038209.2 | 6 | 631 | ||
| NM_008097.2 | NP_032123.3 | 5 | 688 | |||
| GenBank mRNA | AK078866.1 | - | 3 | 241 | ||
| AK165406.1 | - | 5 | 514 | |||
| AU080952.1 | - | 6 | 488 | |||
| BC061158.1 | - | 5 | 518 | 30314273 | ||
| U18992.1 | - | 5 | 469 | |||

Gcdh
Genomic Map