siRNA Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173014.1 | NP_766602.1 | 11 | 1195 | ||
| GenBank mRNA | AB244716.1 | - | 11 | 1090 | ||
| AK039431.1 | - | 11 | 1195 | |||
| BC144805.1 | - | 10 | 1089 | |||
| BC144806.1 | - | 9 | 1030 | 9053354 | ||
| BC150666.1 | - | 9 | 1030 | 9087577 | ||

Lpcat2
Genomic Map