siRNA Details
| Gene Symbol: Rpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016738.5 | NP_058018.2 | 6 | 610 | ||
| GenBank mRNA | AK002787.1 | - | 5 | 547 | ||
| AK010989.1 | - | 6 | 547 | |||
| AK198076.1 | - | 3 | 170 | |||
| AK202315.1 | - | 3 | 170 | |||
| AK204248.1 | - | 3 | 171 | |||
| BC034449.1 | - | 5 | 525 | |||
| BC055358.1 | - | 5 | 523 | 5352468 | ||
| BC083148.1 | - | 3 | 1822 | |||
| BC085493.1 | - | 526 | 6815564 | |||
| CF751804.1 | - | 6 | 663 | |||
| U28917.1 | - | 5 | 528 | |||

Rpl13
Genomic Map