siRNA Details
| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175561.4 | NP_780770.2 | 5 | 1339 | ||
| GenBank mRNA | AK043903.1 | - | 3 | 700 | ||
| BC068235.1 | - | 5 | 1315 | |||
| BC132334.1 | - | 1 | 562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcnxl2| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175561.4 | NP_780770.2 | 5 | 1339 | ||
| GenBank mRNA | AK043903.1 | - | 3 | 700 | ||
| BC068235.1 | - | 5 | 1315 | |||
| BC132334.1 | - | 1 | 562 | |||