siRNA Details
| Gene Symbol: Irf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164598.1 | NP_001158070.1 | 2 | 1338 | ||
| GenBank mRNA | AK087491.1 | - | 1 | 47 | ||
| BC038062.1 | - | 1 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irf2bp2| Gene Symbol: Irf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164598.1 | NP_001158070.1 | 2 | 1338 | ||
| GenBank mRNA | AK087491.1 | - | 1 | 47 | ||
| BC038062.1 | - | 1 | 158 | |||