siRNA Details
| Gene Symbol: Fat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080814.1 | NP_001074283.1 | 20 | 11603 | ||
| GenBank mRNA | AK035028.1 | - | 3 | 408 | ||
| BC117744.1 | - | 3 | 383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat3| Gene Symbol: Fat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080814.1 | NP_001074283.1 | 20 | 11603 | ||
| GenBank mRNA | AK035028.1 | - | 3 | 408 | ||
| BC117744.1 | - | 3 | 383 | |||