siRNA Details
| Gene Symbol: Zbtb44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115130.1 | NP_001108602.1 | 6 | 6398 | ||
| NM_172765.3 | NP_766353.2 | 6 | 6344 | |||
| GenBank mRNA | AK053995.1 | - | 1 | 1823 | ||
| AK153955.1 | - | 1 | 1525 | |||
| BC090965.1 | - | 1 | 1005 | |||

Zbtb44
Genomic Map