siRNA Details
| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081369.2 | NP_001074838.2 | 7 | 586 | ||
| GenBank mRNA | BC038167.1 | - | 5 | 658 | ||
| CX240611.1 | - | 7 | 563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc153| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081369.2 | NP_001074838.2 | 7 | 586 | ||
| GenBank mRNA | BC038167.1 | - | 5 | 658 | ||
| CX240611.1 | - | 7 | 563 | |||