siRNA Details
Gene Symbol: Ccdc153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081369.2 | NP_001074838.2 | 5 | 390 | ||
GenBank mRNA | BC038167.1 | - | 3 | 441 | ||
CX240611.1 | - | 5 | 367 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081369.2 | NP_001074838.2 | 5 | 390 | ||
GenBank mRNA | BC038167.1 | - | 3 | 441 | ||
CX240611.1 | - | 5 | 367 |