siRNA Details
| Gene Symbol: AU019823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134902.1 | NP_001128374.1 | 5 | 558 | ||
| NM_212449.2 | NP_997614.2 | 4 | 531 | |||
| GenBank mRNA | AK133777.1 | - | 4 | 905 | ||
| BC067205.1 | - | 4 | 313 | 6335744 | ||
| BY740784.1 | - | 5 | 375 | |||

AU019823
Genomic Map