siRNA Details
| Gene Symbol: Gm684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195681.1 | NP_001182610.1 | 5 | 645 | ||
| GenBank mRNA | BM935103.1 | - | 5 | 463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm684| Gene Symbol: Gm684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195681.1 | NP_001182610.1 | 5 | 645 | ||
| GenBank mRNA | BM935103.1 | - | 5 | 463 | ||