siRNA Details
| Gene Symbol: Rab39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175562.3 | NP_780771.1 | 2 | 473 | ||
| GenBank mRNA | AB232635.1 | - | 2 | 356 | ||
| AK041076.1 | - | 1 | 50 | |||
| AK049038.1 | - | 2 | 473 | |||
| AK053487.1 | - | 2 | 470 | |||
| BC049787.1 | - | 2 | 453 | 6511404 | ||

Rab39
Genomic Map