siRNA Details
| Gene Symbol: Myo9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173018.2 | NP_766606.2 | 43 | 7645 | ||
| GenBank mRNA | AK028873.1 | - | 13 | 1594 | ||
| AK029836.1 | - | 12 | 1458 | |||
| BC046526.1 | - | 6 | 707 | |||
| BC068298.1 | - | 16 | 2026 | |||
| BC096035.1 | - | 5 | 654 | |||

Myo9a
Genomic Map