siRNA Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173781.4 | NP_776142.1 | 5, 6 | 481 | ||
| GenBank mRNA | AB232597.1 | - | 5, 6 | 398 | ||
| AK035893.1 | - | 5, 6 | 440 | |||
| BC059903.1 | - | 1, 2 | 64 | |||
| BC060618.1 | - | 5, 6 | 481 | 6406134 | ||

Rab6b
Genomic Map