siRNA Details
| Gene Symbol: Gm648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033372.2 | NP_001028544.1 | 4 | 732 | ||
| GenBank mRNA | AK146653.1 | - | 4 | 733 | ||
| BC147598.1 | - | 4 | 732 | 9056588 | ||
| BC147617.1 | - | 4 | 732 | 9056607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm648| Gene Symbol: Gm648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033372.2 | NP_001028544.1 | 4 | 732 | ||
| GenBank mRNA | AK146653.1 | - | 4 | 733 | ||
| BC147598.1 | - | 4 | 732 | 9056588 | ||
| BC147617.1 | - | 4 | 732 | 9056607 | ||