siRNA Details
| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175563.5 | NP_780772.1 | 6 | 778 | ||
| GenBank mRNA | AK040348.1 | - | 6 | 756 | ||
| AK047463.1 | - | 6 | 756 | |||
| AK083651.1 | - | 6 | 773 | |||
| BC023918.1 | - | 6 | 788 | 5323297 | ||

Prr11
Genomic Map