siRNA Details
| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146246.3 | NP_666358.2 | 4 | 2434 | ||
| GenBank mRNA | AJ491325.1 | - | 3 | 2236 | ||
| AY168340.1 | - | 3 | 2215 | |||
| BC129887.1 | - | 4 | 2359 | 40111348 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rp1l1| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146246.3 | NP_666358.2 | 4 | 2434 | ||
| GenBank mRNA | AJ491325.1 | - | 3 | 2236 | ||
| AY168340.1 | - | 3 | 2215 | |||
| BC129887.1 | - | 4 | 2359 | 40111348 | ||