siRNA Details
| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081166.2 | NP_001074635.2 | 13 | 2766 | ||
| GenBank mRNA | AK033659.1 | - | 2 | 1219 | ||
| AK046530.1 | - | 11 | 3021 | |||
| BC044847.1 | - | 1 | 3618 | |||
| BC048859.1 | - | 1 | 3618 | |||
| BC059040.1 | - | 1 | 1586 | |||
| BC067021.1 | - | 13 | 2410 | |||
| BC157925.1 | - | 13 | 2388 | 9088098 | ||
| BC158044.1 | - | 13 | 2388 | 9007348 | ||
| BC172019.1 | - | 13 | 2388 | 9054305 | ||
| BC172145.1 | - | 12 | 2365 | 9054815 | ||

Phf21b
Genomic Map