siRNA Details
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 23 | 2477 | ||
| GenBank mRNA | AB115421.1 | - | 23 | 2472 | ||
| AK140793.1 | - | 23 | 2473 | |||
| BC145012.1 | - | 23 | 2366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13a| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 23 | 2477 | ||
| GenBank mRNA | AB115421.1 | - | 23 | 2472 | ||
| AK140793.1 | - | 23 | 2473 | |||
| BC145012.1 | - | 23 | 2366 | |||