siRNA Details
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 30 | 3291 | ||
| GenBank mRNA | AB115421.1 | - | 30 | 3286 | ||
| AK142462.1 | - | 6 | 505 | |||
| BC145012.1 | - | 30 | 3180 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13a| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 30 | 3291 | ||
| GenBank mRNA | AB115421.1 | - | 30 | 3286 | ||
| AK142462.1 | - | 6 | 505 | |||
| BC145012.1 | - | 30 | 3180 | |||