siRNA Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033373.2 | NP_001028545.2 | 2 | 289 | ||
| GenBank mRNA | BC147314.1 | - | 2 | 289 | 9056364 | |
| BC147315.1 | - | 2 | 289 | 9056370 | ||
| BC147758.1 | - | 2 | 289 | 9087483 | ||
| BC147763.1 | - | 2 | 289 | 9087488 | ||
| BC172036.1 | - | 2 | 289 | 9054322 | ||

Cdk15
Genomic Map