siRNA Details
| Gene Symbol: Rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173031.3 | NP_766619.2 | 7 | 646 | ||
| GenBank mRNA | AK028610.1 | - | 7 | 648 | ||
| BC150940.1 | - | 7 | 601 | |||
| BC172093.1 | - | 7 | 601 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp8nl| Gene Symbol: Rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173031.3 | NP_766619.2 | 7 | 646 | ||
| GenBank mRNA | AK028610.1 | - | 7 | 648 | ||
| BC150940.1 | - | 7 | 601 | |||
| BC172093.1 | - | 7 | 601 | |||