siRNA Details
| Gene Symbol: Lppr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178756.4 | NP_848871.1 | 6 | 856 | ||
| GenBank mRNA | AK034251.1 | - | 6 | 951 | ||
| AK043762.1 | - | 6 | 758 | |||
| AK047614.1 | - | 6 | 786 | |||
| AK087439.1 | - | 6 | 1007 | |||
| AK087514.1 | - | 6 | 857 | |||
| AK160897.1 | - | 6 | 983 | |||
| AY345342.1 | - | 6 | 856 | |||
| BC096762.1 | - | 6 | 832 | 30094664 | ||

Lppr1
Genomic Map