siRNA Details
| Gene Symbol: Irf2bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178757.3 | NP_848872.2 | 1 | 1645 | ||
| GenBank mRNA | AK031834.1 | - | 1 | 1653 | ||
| AK032010.1 | - | 1 | 1590 | |||
| AK083013.1 | - | 1 | 1285 | |||
| AK087457.1 | - | 1 | 1658 | |||
| BC019164.1 | - | 1 | 1326 | 3962494 | ||
| BC036162.1 | - | 1 | 1645 | 4984359 | ||

Irf2bp1
Genomic Map